Learn More
Fisherbrand™ White Caps for Serum Transport Tubes
For 10 mL Polypropylene Transport Tubes
Brand: Fisherbrand™
51.10 EUR valid until 2024-12-31
Use promo code "23291" to get your promotional price.
Additional Details : Weight : 0.01100kg
Specifications
White | |
Polypropylene | |
Nonsterile | |
Transport Tube Caps |