Learn More
Fisherbrand™ Square Labels
GREENER_CHOICE
Fisher branded Cryogenic Storage Labels use a special adhesive that is designed to withstand extreme cold temperatures as low as -196Âş C (-311Âş F). Our cryogenic storage labels can be printed or written on and will not bleed, smudge or smear. Engineered for you to use in your laser printer.
Brand:  Fisherbrand™ 11010212
 View more versions of this product
Product Code. 17837201
  • 58.00€ / 20 sheets
Only null left
Estimated Shipment: 19-08-2025
Description
  • Labels can be printed or written on and will not bleed, smudge or smear
  • Engineered for use in laser printers
Specifications
Specifications
Red
Non-sterile
156/Pk.
Square label sheet
156
0.64 x 3.81cm
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Fisherbrand™ Square Labels > .25 x 1.5 in. (0.64 x 3.81cm); Red; 156 labels/sheet