Learn More
Fisherbrand™ SureOne™ Reload Empty Racks
GREENER_CHOICE
For use with Fisherbrand SureOne universal pipet tips
Brand:  Fisherbrand™ T200ER-FIS
16.35 EUR valid until 2025-06-30
Use promo code "24156" to get your promotional price.
 View more versions of this product
Product Code. 11983456
  • 17.21€ / Pack of 10
Spotlight
Use promo code 24156 at checkout
View Available Promotions
Only null left
In Stock
Edge
Add to basket
Description
Description
  • Inserts (not included) sit flush with rack, allowing easy multichannel pipetting
  • Notched clasp designed to prop lid open for autoclave venting
  • Sure stack lid rim prevents racks from sliding around when stacked and carted around a lab
  • Safe lock latch allows for one-handed opening and closing ability—won't pop open when dropped
  • Sure snap design securely holds insert in place, preventing it from being lifted out while pipetting
  • Flip-lid can be either a hinged or lift-off lid; hinged format does not prevent use of a multichannel pipet
Order Info
Inserts not included; must be ordered separately.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Empty Rack
Pipette Tips
200 to 300 ÎĽL
Videos
  • Talking Points - Fisherbrand Elite Pipettors and SureOne Pipettor Tips (EN)
    Extremely low plunger forces work longer without fatigue and reduce the risk of RSI.
    Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
      Talking Points - Fisherbrand Elite Pipettors
    SDS
    Documents Certificates
    View accessory products
    Promotions
    Promotions
    • Multiple Manufacturers
      Save big on essential products for cancer research
      Promo Code24147
      Expires2025-07-31
      New Offer
      Save Now
    • Multiple Manufacturers
      Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.
      Promo Code: 24156
      Expires2025-06-30
      Days
      30
      Hours
      21
      Minutes
      27
      Seconds
      26
      Hot Deal
      *Discount will apply to qualifying orders received by Fisher Scientific between 01/05/2025 and 30/06/2025. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.
      Terms & Conditions See Full Promo Details
    • Multiple Manufacturers
      Save up to 59% on some of our most popular products
      Promo Code: 24140
      Expires2025-08-24
      Days
      85
      Hours
      21
      Minutes
      27
      Seconds
      26
    Product Content Correction
    Product Title
    Fisherbrand™ SureOne™ Reload Empty Racks > For tip volume: 200 to 300ÎĽL