Learn More
Fisherbrand™ Zipper Seal Sample Bags
Zipper-locking feature securely seals bags
Brand: Fisherbrand™ 9661160
This item is not returnable.
View return policy
Description
Zipper-locking feature securely seals bags
- Durable, transparent polyethylene
- Ideal for collecting, storing, and protecting laboratory specimens and samples easily and conveniently
- Clear, print-free surface lets user mark and label each bag separately
Not suggested for use with liquids
Specifications
Not Autoclavable | |
Zip Seal | |
Polyethylene | |
50 | |
18 x 9 in. (46 x 23cm) | |
46 cm | |
23 cm |
3 lb. | |
Clear | |
250 | |
4 mil | |
18 in. | |
9 in. |
Product Content Correction
Product Title
Fisherbrand™ Zipper Seal Sample Bags > 4mil; 18L x 9 in. W (46 x 23cm)