Learn More
Zeiss™ Fixed Slider
Designed for reflected light with shutter function
Brand: Zeiss™ 428307-9000-000
This item is not returnable.
View return policy
Description
Compatible Microscopes:
- Axio Scope.A1
- Axio Scope.A1 Pol
- Axio Scope.A1 Vario
- Axio Vert.A1 FL
- Axio Vert.A1 MAT
Specifications
Fixed Slider A | |
2 positions for filters with 25mm dia. | |
1 Ea. | |
14 x 40 mm |