Learn More
Zeiss™ Fixed Slider
Designed for reflected light with shutter function
Brand:  Zeiss™ 428307-9000-000
Product Code. 11805930
  • 172.00€ / Each
Estimated Shipment: 04-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Compatible Microscopes:
  • Axio Scope.A1
  • Axio Scope.A1 Pol
  • Axio Scope.A1 Vario
  • Axio Vert.A1 FL
  • Axio Vert.A1 MAT
Specifications
Specifications
Fixed Slider A
2 positions for filters with 25mm dia.
1 Ea.
14 x 40 mm