Learn More
Flash Point Standard, COC ASTM D92 (Nominal Value: 262.0 °C ), Paragon Scientific™
These are certified reference materials characterized by a method specific multi-laboratory study in accordance with ASTM D92. These materials are suitable for verifying the performance of an instrument and/or procedure as specified in this standard.
Brand:  Paragon Scientific CRMU-COC-HIGH
Product Code. 16090030
  • 525.00€ / Pack of 3
Estimated Shipment: 21-01-2025
Description
  • Certified Reference Material for annual verification checks in accordance with ASTM D92.
  • Certified according to ISO 17025 / ISO 17034 under UKAS accreditation by means of a method specific inter-laboratory study.
  • Packaged in 3 x 80 mL bottles – ‘One shot’, one bottle per test ensures zero contamination/ high-end volatile loss.
  • Fully traceable to international standards.
  • Supplied with MSDS.
  • Tamper evident security packaging.
  • 12 month shelf life.
  • Manufactured in the United Kingdom
Specifications
Specifications
3 x 80 mL
Nominal values quoted. Certified Values will vary from batch to batch.