Learn More
Memmert™ Flexible Pt100 Temperature Sensor for UF55plus
Flexible temperature sensor for positioning in chamber or in load.
Brand:  Memmert™ UF55plusH8
Product Code. 17519515
  • 226.00€ / Each
Estimated Shipment: 24-06-2025
Explore more special offers
Add to basket
Description
Description
  • Positioned flexibly in chamber or load
  • For local temperature measurement (up to 1 additional sensor is possible)
  • The measured temperature will be indicated on the display, recorded in the integral data store, and can be documented via the AtmoCONTROL software
  • For Memmert Ovens
Specifications
Specifications
UF55plus Oven
Temperature Sensor
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Memmert™ Flexible Pt100 Temperature Sensor for UF55plus > For Use With: UF55plus