missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FLJ14154 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
198.00€ - 468.00€
Specifications
Antigen | FLJ14154 |
---|---|
Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18361017
|
Bio-Techne
NBP3-03383-20UL |
20 μL |
198.00€
20µL |
Estimated Shipment: 31-01-2025
Log in to see stock. |
|||||
18392224
|
Bio-Techne
NBP3-03383-100UL |
100 μL |
468.00€
100µL |
Estimated Shipment: 31-01-2025
Log in to see stock. |
|||||
Description
FLJ14154 Polyclonal specifically detects FLJ14154 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
FLJ14154 | |
Polyclonal | |
Purified | |
RUO | |
PBS with 50% glycerol, pH7.3. | |
79903 | |
IgG | |
Affinity purified |
Western Blot, Immunohistochemistry, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
EC 2.3.1, EC 2.3.1.-, FLJ11693, FLJ14154, N-acetyltransferase 15, N-acetyltransferase 15 (GCN5-related, putative) | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-242 of human FLJ14154 (NP_079121.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title