Learn More
BD™ Fluid A, Double Wrapped
Packaged in two Stericlin™ bags before autoclaving for a bottle exterior free from microbial contaminants. BD™ Fluid A, Double Wrapped is a diluting and rinsing fluid for sterility tests of pharmaceutical products according to the membrane filtration method.
Brand:  BD™ 257603
Product Code. 15857731
  • 135.00€ / Pack of 10
Edge
Explore more special offers
Description
Description
Fluid A is a diluting and rinsing fluid according to the United States and the European Pharmacopeias.
  • Used for dissolving materials in test and for rinsing sterile devices or materials
  • Releases contaminating organisms from the device or material in test without affecting their viability
  • Proteose Peptone No. 3 acts as a stabilizer for microorganisms, maintaining their viability
  • Terminally sterilized inside a USP infusion bottle with septum for sterile extraction
  • Sterile bottle packaged in two Stericlin™ bags for use in sterile production areas
TRUSTED_SUSTAINABILITY
Specifications
Specifications
USP, EP
Liquid
Sterile
No Polysorbate 80
Per Liter Purified Water: Peptic Digest of Animal Tissue - 1 g
7.1 ± 0.2
Store containers in the dark at 5°-25° C
Diluting/Rinsing Fluid
Autoclaved
Infusion bottles double wrapped in Stericlin™ bags
For dissolving materials and for rinsing devices or materials
100 mL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
BD™ Fluid A, Double Wrapped > Fluid A, Double Wrapped: 100mL