Learn More
Thermo Scientific™ Fluid Transfer Assembly, Adaptor Set
Designed to aid many common shop floor processes such as sampling, transfer, and filling, a fit-for-purpose solution in the required tubing size and length can be easily selected from these designs.
682.00€ - 2160.00€
Product Code | Brand | Tubing Style | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Tubing Style | Price | Quantity & Availability | |||||
17850577
|
Thermo Scientific™
SH31121.01 |
3.22 mm (1/8 in.) ID x 6.35 mm (1/4 in.) OD to 9.52 mm (3/8 in.) ID x 15.88 mm (5/8 in.) OD; TPE 374; 1 m (3.25 ft); Luer to MPC |
2160.00€
Pack of 10 |
Estimated Shipment: 20-01-2025
Log in to see stock. |
|||||
17870647
|
Thermo Scientific™
SH31122.01 |
3.22 mm (1/8 in.) ID x 6.35 mm (1/4 in.) OD to 9.52 mm (3/8 in.) ID x 15.88 mm (5/8 in.) OD; TPE 374; 2 m (6.6 ft); Luer to MPC |
682.00€
Pack of 10 |
Estimated Shipment: 20-01-2025
Log in to see stock. |
|||||
17830577
|
Thermo Scientific™
SH31123.01 |
6.35 mm (1/4 in.) ID x 11.11 mm (7/16 in.) OD to 12.7 mm (1/2 in.) ID x 19.05 mm (3/4 in.) OD; TPE 374; 2 m (6.6 ft); MPX to MPC |
733.00€
Pack of 10 |
Estimated Shipment: 20-01-2025
Log in to see stock. |
|||||
17870577
|
Thermo Scientific™
SH31124.01 |
6.35 mm (1/4 in.) ID x 11.11 mm (7/16 in.) OD to 12.7 mm (1/2 in.) ID x 19.05 mm (3/4 in.) OD; TPE 374; 2 m (6.6 ft); MPX to MPC |
738.00€
Pack of 10 |
Estimated Shipment: 20-01-2025
Log in to see stock. |
|||||
17860577
|
Thermo Scientific™
SH31125.01 |
6.35 mm (1/4 in.) ID x 11.11 mm (7/16 in.) OD to 12.7 mm (1/2 in.) ID x 19.05 mm (3/4 in.) OD; TPE 374; 2 m (6.6 ft); MPX to MPC |
733.00€
Pack of 10 |
Estimated Shipment: 20-01-2025
Log in to see stock. |
|||||
17890647
|
Thermo Scientific™
SH31126.01 |
6.35 mm (1/4 in.) ID x 11.11 mm (7/16 in.) OD to 12.7 mm (1/2 in.) ID x 19.05 mm (3/4 in.) OD; TPE 374; 2 m (6.6 ft); MPX to MPC |
733.00€
Pack of 10 |
Estimated Shipment: 20-01-2025
Log in to see stock. |
|||||
17800587
|
Thermo Scientific™
SH31127.01 |
9.52 mm (3/8 in.) ID x 15.88 mm (5/8 in.) OD to 12.7 mm (1/2 in.) ID x 19.05 mm (3/4 in.) OD; TPE 374; 2 m (6.6 ft); MPX to MPC |
738.00€
Pack of 10 |
Estimated Shipment: 20-01-2025
Log in to see stock. |
|||||
17800657
|
Thermo Scientific™
SH31128.01 |
9.52 mm (3/8 in.) ID x 15.88 mm (5/8 in.) OD to 12.7 mm (1/2 in.) ID x 19.05 mm (3/4 in.) OD; TPE 374; 2 m (6.6 ft); MPX to MPC |
744.00€
Pack of 10 |
Estimated Shipment: 20-01-2025
Log in to see stock. |
|||||
17830657
|
Thermo Scientific™
SH31129.01 |
9.52 mm (3/8 in.) ID x 15.88 mm (5/8 in.) OD to 12.7 mm (1/2 in.) ID x 19.05 mm (3/4 in.) OD; TPE 374; 2 m (6.6 ft); MPX to MPC |
738.00€
Pack of 10 |
Estimated Shipment: 20-01-2025
Log in to see stock. |
|||||
17840657
|
Thermo Scientific™
SH31130.01 |
9.52 mm (3/8 in.) ID x 15.88 mm (5/8 in.) OD to 12.7 mm (1/2 in.) ID x 19.05 mm (3/4 in.) OD; TPE 374; 2 m (6.6 ft); MPX to MPC |
738.00€
Pack of 10 |
Estimated Shipment: 20-01-2025
Log in to see stock. |
|||||
Description
Standard Fluid Transfer Assembly products interconnect with Thermo Scientific standard Labtainer BPCs, Productainer BPCs, single-use bioreactors, mixers, and fermentors right on the shop floor to deliver a comprehensive and complete set of single-use products to meet the needs of your facility or process.- One set of stocked products reduces the number of customized stock keeping units (SKUs), simplifies the ordering process, and reduces product storage requirements
- Multiple manufacturing sites ensure security in product supply
- Produced under cGMP conditions in ISO 7 clean areas to ensure a high level of quality
- Characterized using ISO, USP, EP, and ASTM test methods to validate performance
Specifications
Adapter Set |