Learn More
Thermo Scientific™ Fluid Transfer Assembly, Extension Set
Designed to aid many common shop floor processes such as sampling, transfer, and filling, a fit-for-purpose solution in the required tubing size and length can be easily selected from these designs.
315.00€ - 2140.00€
Product Code | Brand | Tubing Style | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Tubing Style | Price | Quantity & Availability | |||||
17850687
|
Thermo Scientific™
SH31116.01 |
9.52 mm (3/8 in.) ID; 15.88 mm (5/8 in.) OD; TPE 374; 1 m (3.25 ft); MPC |
1660.00€
Pack of 10 |
Estimated Shipment: 20-01-2025
Log in to see stock. |
|||||
17880687
|
Thermo Scientific™
SH31116.02 |
9.52 mm (3/8 in.) ID; 15.88 mm (5/8 in.) OD; TPE 374; 2.5 m (8.2 ft); MPC |
672.00€
Pack of 10 |
Estimated Shipment: 20-01-2025
Log in to see stock. |
|||||
17860637
|
Thermo Scientific™
SH31116.03 |
9.52 mm (3/8 in.) ID; 15.88 mm (5/8 in.) OD; TPE 374; 5 m (16.4 ft); MPC |
1970.00€
Pack of 10 |
Estimated Shipment: 20-01-2025
Log in to see stock. |
|||||
17810697
|
Thermo Scientific™
SH31117.01 |
6.35 mm (1/4 in.) ID; 11.11 mm (7/16 in.) OD; TPE 374; 1 m (3.25 ft); Luer |
1425.00€
Pack of 10 |
Estimated Shipment: 20-01-2025
Log in to see stock. |
|||||
17890687
|
Thermo Scientific™
SH31117.02 |
6.35 mm (1/4 in.) ID; 11.11 mm (7/16 in.) OD; TPE 374; 2.5 m (8.2 ft); Luer |
641.00€
Pack of 10 |
Estimated Shipment: 20-01-2025
Log in to see stock. |
|||||
17800697
|
Thermo Scientific™
SH31117.03 |
6.35 mm (1/4 in.) ID; 11.11 mm (7/16 in.) OD; TPE 374; 5 m (16.4 ft); Luer |
1845.00€
Pack of 10 |
Estimated Shipment: 20-01-2025
Log in to see stock. |
|||||
17820697
|
Thermo Scientific™
SH31118.01 |
6.35 mm (1/4 in.) ID; 11.11 mm (7/16 in.) OD; TPE 374; 1 m (3.25 ft); MPC |
346.00€
Pack of 10 |
Estimated Shipment: 20-01-2025
Log in to see stock. |
|||||
17830647
|
Thermo Scientific™
SH31118.02 |
6.35 mm (1/4 in.) ID; 11.11 mm (7/16 in.) OD; TPE 374; 2.5 m (8.2 ft); MPC |
315.00€
Pack of 10 |
Estimated Shipment: 20-01-2025
Log in to see stock. |
|||||
17800647
|
Thermo Scientific™
SH31118.03 |
6.35 mm (1/4 in.) ID; 11.11 mm (7/16 in.) OD; TPE 374; 5 m (16.4 ft); MPC |
995.00€
Pack of 10 |
Estimated Shipment: 20-01-2025
Log in to see stock. |
|||||
17890567
|
Thermo Scientific™
SH31119.01 |
3.22 mm (1/8 in.) ID; 6.35 mm (1/4 in.) OD; TPE 374; 1 m (3.25 ft); Luer |
1395.00€
Pack of 10 |
Estimated Shipment: 20-01-2025
Log in to see stock. |
|||||
17820647
|
Thermo Scientific™
SH31119.02 |
3.22 mm (1/8 in.) ID; 6.35 mm (1/4 in.) OD; TPE 374; 2.5 m (8.2 ft); Luer |
564.00€
Pack of 10 |
Estimated Shipment: 20-01-2025
Log in to see stock. |
|||||
17840647
|
Thermo Scientific™
SH31119.03 |
3.22 mm (1/8 in.) ID; 6.35 mm (1/4 in.) OD; TPE 374; 5 m (16.4 ft); Luer |
1720.00€
Pack of 10 |
Estimated Shipment: 20-01-2025
Log in to see stock. |
|||||
17850647
|
Thermo Scientific™
SH31120.01 |
12.7 mm (1/2 in.) ID; 19.05 mm (3/4 in.) OD; TPE 374; 1 m (3.25 ft); MPX |
1795.00€
Pack of 10 |
Estimated Shipment: 20-01-2025
Log in to see stock. |
|||||
17860647
|
Thermo Scientific™
SH31120.02 |
12.7 mm (1/2 in.) ID; 19.05 mm (3/4 in.) OD; TPE 374; 2.5 m (8.2 ft); MPX |
867.00€
Pack of 10 |
Estimated Shipment: 20-01-2025
Log in to see stock. |
|||||
17810577
|
Thermo Scientific™
SH31120.03 |
12.7 mm (1/2 in.) ID; 19.05 mm (3/4 in.) OD; TPE 374; 5 m (16.4 ft); MPX |
2140.00€
Pack of 10 |
Estimated Shipment: 20-01-2025
Log in to see stock. |
|||||
Description
Standard Fluid Transfer Assembly products interconnect with Thermo Scientific standard Labtainer BPCs, Productainer BPCs, single-use bioreactors, mixers, and fermentors right on the shop floor to deliver a comprehensive and complete set of single-use products to meet the needs of your facility or process.- One set of stocked products reduces the number of customized stock keeping units (SKUs), simplifies the ordering process, and reduces product storage requirements
- Multiple manufacturing sites ensure security in product supply
- Produced under cGMP conditions in ISO 7 clean areas to ensure a high level of quality
- Characterized using ISO, USP, EP, and ASTM test methods to validate performance
- Sterility: Irradiation dose of 25-40 kGy provides minimum SAL of 10-6 for product contact surfaces
Specifications
Extension Set |