missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fluorescein Antibody (4-4-20 (enhanced)), Allophycocyanin, Novus Biologicals™
Human Monoclonal Antibody
Brand: Bio-Techne NBP2-81024APC
This item is not returnable.
View return policy
Description
Fluorescein Monoclonal specifically detects Fluorescein in Non-species specific samples. It is validated for Flow Cytometry.
Specifications
Fluorescein | |
Monoclonal | |
APC | |
PBS | |
Human | |
Protein A purified | |
RUO | |
Fluorescein labelled proteins. | |
Store at 4°C in the dark. | |
IgG1 Îş |
Flow Cytometry | |
4-4-20 (enhanced) | |
Flow Cytometry | |
FITC, fluorescein isothyocianate | |
Fluorescein I-bovine serum albumin (FlI-BSA). | |
0.1 mL | |
Primary | |
Non-species specific | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fluorescein Antibody (4-4-20 (enhanced)), Allophycocyanin, Novus Biologicals™ > 0.1 mL; APC
Spot an opportunity for improvement?Share a Content Correction