Learn More
Fluquinconazole, Honeywell Fluka™
Brand:  Honeywell Fluka R1688-100MG
Product Code. 16676133
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »
Explore more special offers
Description
Description
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials  and organic reference materials.
  • Produced according to ISO 9001
  • Suitable for a wide range of industries and applications
  • Detailed Certificate of Analysis included with every delivery
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Fluquinconazole
≤ 100%
376.17 g/mol
1.58 g/cm3 (20°C)
White
136426-54-5
3-(2,4-Dichlorophenyl)-6-fluoro-2-(1H-1,2,4-trizol-1-yl)quinazolin-4(3H)-one
Fungicides and Bactericides
191°C to 193°C