missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Flurprimidol

Product Code. 17201316
Click to view available options
Quantity:
100 mg
500 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56425-91-3
Molecular Formula C15H15F3N2O2

Specifications

Quantity 500 mg
Solubility Solubility aq. soln.: insoluble.
Melting Point 93.0°C to 97.0°C
Boiling Point 264.0°C
Formula Weight 312.29
InChI Key VEVZCONIUDBCDC-UHFFFAOYNA-N
SMILES CC(C)C(O)(C1=CC=C(OC(F)(F)F)C=C1)C1=CN=CN=C1
IUPAC Name 2-methyl-1-(pyrimidin-5-yl)-1-[4-(trifluoromethoxy)phenyl]propan-1-ol
Molecular Weight (g/mol) 312.29
Chemical Name or Material Flurprimidol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'