missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Flurprimidol
Click to view available options
Quantity:
100 mg
500 mg
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Quantity | 100 mg |
Solubility | Solubility aq. soln.: insoluble. |
Melting Point | 93.0°C to 97.0°C |
Boiling Point | 264.0°C |
Formula Weight | 312.29 |
InChI Key | VEVZCONIUDBCDC-UHFFFAOYNA-N |
SMILES | CC(C)C(O)(C1=CC=C(OC(F)(F)F)C=C1)C1=CN=CN=C1 |
IUPAC Name | 2-methyl-1-(pyrimidin-5-yl)-1-[4-(trifluoromethoxy)phenyl]propan-1-ol |
Molecular Weight (g/mol) | 312.29 |
Chemical Name or Material | Flurprimidol |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'