Learn More
ZVG Zellstoff Vertrieb™ Folded Towels
Use ZVG Zellstoff Vertrieb™ Folded Towels in either 2-ply or 3-ply folded paper forms. They are available in various sizes, quantities and colors as well.
Brand: ZVG Zellstoff Vertrieb™ 1542204
33.54 EUR valid until 2025-03-31
Use promo code "24146" to get your promotional price.
This item is not returnable.
View return policy
Description
- ZetForm™l paper towel system
- Various colors
- Various thicknesses
Specifications
Folded Towel | |
21 cm | |
25 x 21 cm |
25 cm | |
Paper | |
2-ply |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 12
Special Offers
-
HC Innovations
,
Vernacare
,
BM Polyco
,
ZVG Zellstoff Vertrieb
,
Fisherbrand
Essential Lab Wipes and Paper Sale
Up to 60% Off on Top-Quality AlternativesPromo Code: 24146Expires: 2025-03-31
Product Content Correction
Product Title
ZVG Zellstoff Vertrieb™ Folded Towels > Thickness: 2-ply; Dimensions: 25L x 21cmW