Specifications
Forceps | |
Stainless Steel | |
For Tissue | |
12 cm |
Product Suggestions
Customers who viewed this item also viewed.
Product Content Correction
Product Title
Rogo Sampaic™ Forceps > Forceps; For Use With Tissue 12cm