Learn More
Labconco™ Four-Receptable Power Strip
Provide electrical receptacles for connecting instruments and equipment
459.00€
Specifications
For Use With (Application) | To provide electrical receptacles for connecting instruments and equipment |
---|---|
Weight (English) | 1 lb. |
Weight (Metric) | 0.5 kg |
Product Code | Brand | Amperage | Includes | Product Type | Voltage | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Amperage | Includes | Product Type | Voltage | Price | Quantity & Availability | |||
11994809
|
Labconco™
5290400 |
2 A | 4 receptacles | European Power Strip | 250 V |
459.00€
Each |
Estimated Shipment: 14-05-2025
Log in to see stock. |
|||
Specifications
To provide electrical receptacles for connecting instruments and equipment | |
1 lb. | |
0.5 kg |
Product Title
Labconco™ Four-Receptable Power Strip