missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOXE1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
198.00€ - 468.00€
Specifications
Antigen | FOXE1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18392456
|
Bio-Techne
NBP3-04805-20UL |
20 μL |
198.00€
20µL |
Estimated Shipment: 31-01-2025
Log in to see stock. |
|||||
18320087
|
Bio-Techne
NBP3-04805-100UL |
100 μL |
468.00€
100µL |
Estimated Shipment: 31-01-2025
Log in to see stock. |
|||||
Description
FOXE1 Polyclonal specifically detects FOXE1 in Human samples. It is validated for Western Blot.Specifications
FOXE1 | |
Polyclonal | |
Purified | |
RUO | |
PBS with 50% glycerol, pH7.3. | |
2304 | |
IgG | |
Affinity purified |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
FKHL15HFKL5, forkhead box E1 (thyroid transcription factor 2), forkhead box E2, Forkhead box protein E2, forkhead, drosophila, homolog-like 15, Forkhead-related protein FKHL15, HFKH4Thyroid transcription factor 2, TITF2HNF-3/fork head-like protein 5, TTF2forkhead box protein E1, TTF-2FOXE2 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 184-373 of human FOXE1 (NP_004464.2). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title