Learn More
VACUUBRAND™ FPM Spear Sealing Kit
Brand: VACUUBRAND™ 20660136
8.36 EUR valid until 2025-04-13
Use promo code "24122" to get your promotional price.
This item is not returnable.
View return policy
Description
Specifications
25 x 28 x 5mm | |
FPM | |
Sealing Ring |
For Small Flanges KF | |
1 Each |
Product Suggestions
Customers who viewed this item also viewed
Special Offers
-
Multiple Manufacturers
Precision at Your Fingertips
Master liquid handling with ease.Promo Code: 24122Expires: 2025-04-13
Days
8Hours
8Minutes
22Seconds
36
Product Content Correction
Product Title
VACUUBRAND™ FPM Spear Sealing Kit > Dimensions: 25 x 28 x 5mm