Learn More
Eppendorf™ Freezer Rack: Innova™ U360, U535, U725/-G, CryoCube™ F740 series (3-comp., Std),
Comprehensive storage of samples in a ULT freezer is based on storage boxes. The boxes should be stored in dedicated freezer racks. These metal racks enable optimal use of space as well as easy relocation of sample boxes.
Brand:  Eppendorf™ 6001011410
Product Code. 15733235
  • 151.60€ / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Freezer Rack: Innova™ U360, U535, U725, U725-G, CryoCube™ F740 series (3-compartment, Standard), 4 in/102 mm, side access, stainless steel- Rack for 102 mm/4 in boxes- Max. box size [mm]: 136 x 136- Side access rack- Ext. dimensions of rack (DxWxH) [mm]: 569 x 139 x 318 (22.4 x 5.5 x 12.5 in)- Max. box number per rack: 12 (4 x 3)- Stainless steel- For ULT freezer: Innova U360 (max. 9 racks), Innova U535 (max. 12 racks), Innova U725 and U725-G (max. 18 racks), CryoCube F740 series (3-compartment version; max. 18 racks)- For ULT freezer: Innova U360 (max. 108 boxes), Innova U535 (max. 144 boxes), Innova U725 and U725-G (max. 216 boxes), CryoCube F740 series (3-compartment version; max. 216 boxes)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Freezer Rack
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products