Learn More
Motic™ FS01 Head Holder (Arms)
Suitable for SFC-11 microscope heads. Motic™ FS01 Head Holder (Arms) is designed with a mount rack and pinion focusing device.
Brand: Motic™ 11143131
This item is not returnable.
View return policy
Description
- FS01 head holder without illumination.
- For 25mm OD pole.
- For 74mm OD head.
- Mount rack and pinion focusing device.
Specifications
Head Holder Without Illumination | |
For 25mm OD pole, 74mm OD head. | |
Mount rack and pinion focusing device |
SFC-11 Microscope Heads | |
SFC-11 microscope heads |