Learn More
3M™ Full Facepiece Reusable Respirator
3M™ Promask™ Full Facepiece Reusable Respirator FM3 is a multi-function full-face mask designed to provide durable and comfortable protection against a wide range of respiratory hazards. Use in conjunction with approved 3M powered air systems or as a negative pressure respirator.
915.00€
Specifications
Connection Type | Bayonet & DIN |
---|---|
Color | Black |
Product Type | Full Face Mask |
Suspension Type | 5 Point Strap |
Product Code | Brand | Size | Model | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Model | Price | Quantity & Availability | |||||
17786973
|
3M™
7100237033 |
Small | FF-301 |
915.00€
Pack of 4 |
Estimated Shipment: 16-07-2025
Log in to see stock. |
|||||
17766973
|
3M™
7100236989 |
Large | FF-302 |
915.00€
Pack of 4 |
Estimated Shipment: 16-07-2025
Log in to see stock. |
|||||
Description
- Use as a negative-pressure single-filter mask or as a face piece for a powered air respirator
- Sweat port helps to provide user comfort during long periods of use
- Visor provides a good field of view and comes in choice of polycarbonate or high-heat material
- Side-fitting filter port and clear inner mask for good forwards and downwards vision
- Filter doesn't interfere with front-mounted speech diaphragm to aid in easy communication
Specifications
Bayonet & DIN | |
Black | |
Full Face Mask | |
5 Point Strap |
Product Title
3M™ Full Facepiece Reusable Respirator