Learn More
Memmert™ Full-Sight Glass Door for UF55plus, Max. 250°C
Full-sight glass door constructed of four insulated glass layers.
Brand: Memmert™ UF55plusB0
This item is not returnable.
View return policy
Description
- Accessory
- Full-sight
- Glass (4 layers)
- Temperature range up to 250°C
Specifications
UF55plus Oven | |
250°C (Max.) | |
Full-sight Door | |
Glass |
Product Suggestions
Customers who viewed this item also viewed.
Product Content Correction
Product Title
Memmert™ Full-Sight Glass Door for UF55plus, Max. 250°C > For Use With: UF55plus