missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Fulvestrant

Product Code. 16627042
Click to view available options
Quantity:
50 mg
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 129453-61-8
Molecular Formula C32 H47 F5 O3 S

Specifications

Quantity 50 mg
Solubility (0.1 g in 10 mL Ethanol) Clear solution
Formula Weight 606.77
Physical Form Powder
Product Type Reagent
Purity ≥98.5% (HPLC)
InChI Key VWUXBMIQPBEWFH-WCCTWKNTSA-N
SMILES C[C@]12CC[C@H]3[C@H]([C@@H]1CC[C@@H]2O)[C@H](CCCCCCCCCS(=O)CCCC(F)(F)C(F)(F)F)CC1=CC(O)=CC=C31
IUPAC Name (1S,3aS,3bR,4R,9bS,11aS)-11a-methyl-4-[9-(4,4,5,5,5-pentafluoropentanesulfinyl)nonyl]-1H,2H,3H,3aH,3bH,4H,5H,9bH,10H,11H,11aH-cyclopenta[a]phenanthrene-1,7-diol
Molecular Weight (g/mol) 606.78
Chemical Name or Material Fulvestrant
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'