missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GAB1 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
198.00€ - 468.00€
Specifications
Antigen | GAB1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18374575
|
Bio-Techne
NBP3-04454-20UL |
20 μL |
198.00€
20µL |
Estimated Shipment: 31-01-2025
Log in to see stock. |
|||||
18392185
|
Bio-Techne
NBP3-04454-100UL |
100 μL |
468.00€
100µL |
Estimated Shipment: 31-01-2025
Log in to see stock. |
|||||
Description
GAB1 Polyclonal specifically detects GAB1 in Mouse samples. It is validated for Western Blot.Specifications
GAB1 | |
Polyclonal | |
Purified | |
RUO | |
Mouse | |
GRB2-associated binder 1, GRB2-associated binding protein 1, GRB2-associated-binding protein 1, Growth factor receptor bound protein 2-associated protein 1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-150 of human GAB1 (NP_997006.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
Rabbit | |
Apoptosis, Phospho Specific, Signal Transduction, Tyrosine Kinases | |
PBS with 50% glycerol, pH7.3. | |
2549 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title