Learn More
Thermo Scientific Chemicals Galanin, Human
For cellular and molecular biology applications
152.00€ - 271.00€
Specifications
Species | Human |
---|---|
Purification Method | HPLC |
Content And Storage | -30°C to -10°C |
Format | Powder |
Molecular Weight (g/mol) | 3157.41 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15417539
|
Thermo Scientific Chemicals
J66251.!D |
0.5 mg |
152.00€
0.50mg |
Estimated Shipment: 23-01-2025
Log in to see stock. |
|||||
15427539
|
Thermo Scientific Chemicals
J66251.M^ |
1 mg |
271.00€
1mg |
Estimated Shipment: 23-01-2025
Log in to see stock. |
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Human | |
-30°C to -10°C | |
3157.41 | |
RUO - research use only |
HPLC | |
Powder | |
Galanin, human | |
Gly-Trp-Thr-Leu-Asn-Ser-Ala-Gly-Tyr-Leu-Leu-Gly-Pro-His-Ala-Val-Gly-Asn-His-Arg-Ser-Phe-Ser-Asp-Lys-Asn-Gly-Leu-Thr-Ser |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.