Learn More
Ansell GAMMEX™ Neoprene Surgical Glove
Product Code. 17754959
Click to view available options
Size:
5.5
6
6.5
7
7.5
8
8.5
9
Neoprene and chemical accelerator-free surgical gloves that offer unsurpassed combination of sensitivity and durability.
  • Non-latex and 100% chemical accelerator-free
  • Featuring SENSOPRENE™ Technology to deliver a thinner yet robust glove for added comfort and durability
  • Up to 30% thinner on average than Ansell's standard neoprene surgical gloves
  • Smooth with a micro-textured finish for secure instrument handling and grip
  • Tested for use with chemotherapy drugs and a wide range of chemicals
Specifications
AQL 0.65
Color Natural
Disposable Single-use
Exterior Finish Fully Textured
Latex Free Yes
Material Neoprene
Powdered Powder Free
Size 7.5
Sterility Sterile