Learn More
Novus Biologicals™ GAPDH Recombinant Protein Antigen
Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition
Brand: Novus Biologicals™ NBP2-59025PEP
This item is not returnable.
View return policy
Description
A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human GAPDH. The GAPDH Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.Specifications
2597 | |
>80% by SDS-PAGE and Coomassie blue staining | |
PBS and 1M Urea, pH 7.4. | |
aging-associated gene 9 protein, EC 1.2.1, EC 1.2.1.12, EC 2.6.99.-, G3PD, GAPD, glyceraldehyde 3-phosphate dehydrogenase, glyceraldehyde-3-phosphate dehydrogenase, MGC88685, Peptidyl-cysteine S-nitrosylase GAPDH | |
24 kDa | |
RUO | |
This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP2-54336. It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/ml |
Human | |
Store at -20°C. Avoid freeze-thaw cycles. | |
Antibody Competition | |
GAPDH | |
100 μl | |
E. coli |