Learn More
Garcin™ Soap Dispenser
Brand:  Garcin LB921480
Product Code. 11713584
  • 56.00€ / Each
Currently Unavailable: We have no information when this product will be available
View alternative products
Specifications
Soap Dispenser
1000 mL
Product Title
Garcin™ Soap Dispenser > Capacity: 1000mL