Learn More
Thermo Scientific™ Gas Chromatography 3-Port SilFlow Replacement Parts
Switch your GC column in GC-MS without having to vent.
Brand: Thermo Scientific™ 29063467
72.52 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.
Description
Maintain reliability with 3-Port SilFlow replacement parts when you place your next order.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Ferrule | |
SilFlow fingertight ferrules | |
10 Pack | |
for use with columns of OD 0.45 mm (ID 0.32mm) |
Product Suggestions
Customers who viewed this item also viewed
Special Offers
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
Days
26Hours
12Minutes
18Seconds
47 -
Multiple Manufacturers
Chromatography and Chemicals Promotion
Explore our new promotion and get up to 70% off some of our most popular products.Promo Code: 24127Expires: 2025-05-31
Days
57Hours
12Minutes
18Seconds
47New Offer Save Up to 70%