missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GAS2 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
198.00€ - 468.00€
Specifications
Antigen | GAS2 |
---|---|
Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18393387
|
Bio-Techne
NBP3-04395-20UL |
20 μL |
198.00€
20µL |
Estimated Shipment: 31-01-2025
Log in to see stock. |
|||||
18392524
|
Bio-Techne
NBP3-04395-100UL |
100 μL |
468.00€
100µL |
Estimated Shipment: 31-01-2025
Log in to see stock. |
|||||
Description
GAS2 Polyclonal specifically detects GAS2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
GAS2 | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse | |
GAS-2, growth arrest-specific 2, growth arrest-specific protein 2, MGC32610 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-100 of human GAS2 (NP_005247.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Apoptosis | |
PBS with 50% glycerol, pH7.3. | |
2620 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title