Learn More
Product Code | Brand | Material | For Use With (Application) | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Material | For Use With (Application) | Quantity | Price | Quantity & Availability | |||
11924362
|
RETSCH
22.085.0007 |
Stainless Steel, PTFE | For 1.5mL grinding jar | 10 Pack | N/A | ||||
15350984
|
RETSCH
03.111.0313 |
Stainless Steel, PTFE | For 10mL grinding jar | 1 Pack |
9.85€
Each |
Estimated Shipment: 14-03-2025
Log in to see stock. |
|||
10466739
|
RETSCH
22.085.0009 |
Stainless Steel, PTFE | For 10mL grinding jar | 10 Pack | N/A | ||||
15330984
|
RETSCH
03.111.0291 |
Stainless Steel, PTFE | For 25mL grinding jar | 1 Pack |
19.20€
Each |
|
|||
15340984
|
RETSCH
03.111.0296 |
Zirconium Oxide | For 25mL grinding jar | 1 Pack |
19.20€
Each |
|
|||
15320984
|
RETSCH
03.111.0290 |
Stainless Steel, PTFE | For 35mL grinding jar | 1 Pack |
19.20€
Each |
Estimated Shipment: 12-02-2025
Log in to see stock. |
|||
10250315
|
RETSCH
22.085.0003 |
Zirconium Oxide, Tungsten Carbide | For 25mL grinding jar | 10 Pack | N/A | ||||
11944362
|
RETSCH
22.085.0005 |
Stainless Steel | For 35mL grinding jar | 10 Pack | N/A | ||||
15310984
|
RETSCH
03.111.0289 |
Stainless Steel, PTFE | For 50mL grinding jar | 1 Pack |
19.20€
Each |
|
|||
10472080
|
RETSCH
22.085.0004 |
Zirconium Oxide | For 35mL grinding jar | 10 Pack | N/A | ||||
10704787
|
RETSCH
22.085.0002 |
Stainless Steel | For 50mL grinding jar | 10 Pack |
71.45€
Pack of 10 |
|
|||
15300984
|
RETSCH
03.111.0262 |
Stainless Steel, PTFE | For 5mL grinding jar | 1 Pack |
10.55€
Each |
|
|||
11934362
|
RETSCH
22.085.0008 |
Stainless Steel, PTFE | For 5mL grinding jar | 10 Pack | N/A | ||||
10290773
|
RETSCH
22.085.0006 |
Stainless Steel | For 35mL grinding jar | 10 Pack | N/A | ||||
Specifications
Gasket |