Learn More
Heidolph Instruments Gasket Seal for Rotary Evaporators
Made of glass materials, the ultra resistant FFKM Gasket Seal for applications with highly abrasive solutions is ideal in research, scientific, and other laboratory environments.
Brand:  Heidolph Instruments 23-30-01-01-30
65.84 EUR valid until 2025-06-30
Use promo code "24156" to get your promotional price.
Product Code. 10592672
  • 70.90€ / Each
In Stock
Edge
Description
  • Vacuum seal
  • Standard FKM spare seal
  • Ultra resistant FFKM seal for applications with highly abrasive solutions
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PTFE
Gasket
115/230 VAC
Special Offers
  • Multiple Manufacturers
    Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.
    Promo Code: 24156
    Expires2025-06-30
    Days
    53
    Hours
    2
    Minutes
    39
    Seconds
    21
    Hot Deal
    *Discount will apply to qualifying orders received by Fisher Scientific between 01/05/2025 and 30/06/2025. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.
    Terms & Conditions See Full Promo Details
Product Content Correction
Product Title
Heidolph Instruments Gasket Seal for Rotary Evaporators > Gasket Seal for Rotary Evaporators; Material: Glass