Learn More
Thermo Scientific™ GC Liner Sealing Rings for Thermo Scientific™ and Agilent Instruments
Match the best sealing ring with the injection port liner used in your Thermo Scientific™ or Agilent GC system.
Brand: Thermo Scientific™ 29033406
106.92 EUR valid until 2025-04-30
Use promo code "24211" to get your promotional price.
Description
Match the best sealing ring with the injection port liner used in your Thermo Scientific™ or Agilent GC system.
Specifications
Liner Sealing Ring | |
10 Pack | |
Liner Sealing Ring | |
TRACE and FOCUS GC SSL Injectors |
Product Suggestions
Customers who viewed this item also viewed
Special Offers
-
Multiple Manufacturers
Chromatography Monthly Promotion
Discover exclusive monthly deals on chromatography supplies to boost your lab's efficiency.Promo Code: 24211Expires: 2025-04-30
Days
26Hours
9Minutes
16Seconds
54New Offer Save Up to 67% -
Multiple Manufacturers
Chromatography and Chemicals Promotion
Explore our new promotion and get up to 70% off some of our most popular products.Promo Code: 24127Expires: 2025-05-31
Days
57Hours
9Minutes
16Seconds
54New Offer Save Up to 70%