Learn More
Biodesign™ Gel Protector Sheet Gelfile
Brand: Biodesign™ G309
This item is not returnable.
View return policy
Description
Stores, organises and displays dried polyacrylamide and agarose gels- Each BioDesign GelFile™ binder contains heavy crystal clear vinyl pages with pockets and white backgrounds
- The pages are chemically neutral and are guaranteed not to alter your dried gels or turn yellow for TEN years
- Dried gels are held flat for long term storage without curling
- Shields gels from moisture, accidental spills and possible damage
- White background allows optimal comparison between gels
- Each BioDesign GelFile™ binder holds 20 to 25 pages
- A clear label holder on the spine allows you to personalise each binder
Specifications
11.5 x 18 cm | |
Gel Protector Sheets | |
40 Pages (Stores 160 Gels) |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Product Title
Biodesign™ Gel Protector Sheet Gelfile > for gels up to 115mm x 180mm heavy crystal clear vinyl with white background insert BioDesign