Learn More
Corning™ General Assay Microplate Lids
Manufactured with rigid polystyrene for optimum visibility and strength. Corning™ General Assay Microplate Lids are available in a variety of styles and types.
Brand: Corning™ 3098
240.86 EUR valid until 2025-04-13
Use promo code "24122" to get your promotional price.
Description
Specifications
Polystyrene | |
100/Cs. | |
Universal Lid | |
No |
Sterile | |
25/Pk. | |
All Microplates |
Product Suggestions
Customers who viewed this item also viewed
Special Offers
-
Multiple Manufacturers
Precision at Your Fingertips
Master liquid handling with ease.Promo Code: 24122Expires: 2025-04-13
Days
8Hours
15Minutes
52Seconds
59 -
Multiple Manufacturers
Back to the Lab Summer Semester
Save up to 94% on Teaching Lab EssentialsPromo Code: 25721Expires: 2025-05-01
New Offer Save Now -
Corning Matrigel
,
Corning Biocoat
,
Corning CoolProduct
,
Corning Media Tech
,
Corning