Learn More
Product Code | Brand | Includes | Warranty | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Warranty | Price | Quantity & Availability | ||||
17142115
|
Thermo Scientific™
840-285300 |
Performance Verification Kit | Standard |
861.00€
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Get high precision and functionality in a simple yet sophisticated visible spectrophotometer. The Thermo Scientific™ GENESYS™ 30 Visible Spectrophotometer performs both basic and complex analyses for educational or industrial use. Its intuitive design allows students and teachers to focus on scientific data and concepts rather than on learning how to use the instrument. In the lab, technicians of all levels working in routine QA/QC applications can easily obtain credible results to confirm quality or inform next steps. Results can be reported quickly as digital or printed data.
The GENESYS 30 visible spectrophotometer brings you intuitive design, a high definition color screen, fast and accurate measurements, and convenient data reporting. It is the ideal instrument for any user, whether a novice student or an experienced lab associate.
Product Features:
- Entirely new industrial design for better usability
- High definition 5-inch color screen with rubberized push-button keypad for easy navigation
- Single cell measurement with removable, washable sample compartment liner
- Always-on light source for rapid analysis, easy maintenance
- Standard cell holder for 10mm cuvettes and optional holders for other cell types including test-tubes
- New onboard software UI; Thermo Scientific™ VISIONlite PC software is optional
- Data any way you want it
- USB data export in a text format that’s easy to read and process
- Excellent snap-on thermal printer available
- Three-year warranty and skilled technical support
Meets curriculum topics for:
- Single wavelength
- Scanning
- Beer’s Law
- Colorimetry/colorimetric analysis
- Quantitative analysis
Lab associate benefits:
- Higher data confidence with 6-point quant plus concentration mode
- Multiple local language choices