Learn More
Product Code | Brand | Description | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | For Use With (Equipment) | Price | Quantity & Availability | |||||
15841148
|
Thermo Scientific™
840-305000 |
Standard Fiber Optic Dip Probe with 10 mm Tip | GENESYS 150 and GENESYS 180 Spectrophotometers |
3195.00€
Each |
Estimated Shipment: 17-04-2025
Log in to see stock. |
|||||
15896095
|
Thermo Scientific™
840-305700 |
Complete 6 mm Fiber Optic Dip Probe system
Note: Photometric performance of fiber probe systems is different from cuvette systems and may be impacted by ambient lighting conditions |
GENESYS 150 and GENESYS 180 Spectrophotometers |
4760.00€
Each |
Estimated Shipment: 17-04-2025
Log in to see stock. |
|||||
15806105
|
Thermo Scientific™
840-305800 |
Fiber Optic Coupler for GENESYS 150 and GENESYS 180 Spectrophotometers | GENESYS 150 and GENESYS 180 Spectrophotometers |
2235.00€
Each |
Estimated Shipment: 17-04-2025
Log in to see stock. |
|||||
15861148
|
Thermo Scientific™
840-307400 |
10 mm Pathlength Tip for Dip Probe for GENESYS 150 and GENESYS 180 Spectrophotometers | GENESYS 150 and GENESYS 180 spectrophotometers |
1070.00€
Each |
Estimated Shipment: 17-04-2025
Log in to see stock. |
|||||
Description
Dip, tap and measure - eliminate liquid transfer and cuvettes from your workflow.
Preserve sample integrity while minimizing sample carryover.Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Thermo Scientific™ GENESYS Fiber Optic Probes