Learn More
Herka™ Glass Buret
Brand:  Herka™ A1824108
Product Code. 11920567
  • 284.00€ / Each
Estimated Shipment: 05-05-2025
Specifications
2 mL
Glass
0.01 mL
±0.01 mL
Product Title
Herka™ Glass Buret > Capacity: 2mL