man
Test
Search
Order Status
Quick Order
Support
Sign In
chevron-down
chevron-down
Learn More
Home
Products
Facility Safety and Maintenance
Waste Disposal
Biohazardous and Sharps Disposal Containers
11821103
Print
Glass Cardboard
Capacity: 50L
Brand:
Hospidex ONDHOSPI50V
Product Code.
11821103
49.20€
/ Pack of 10
Quantity
In Stock
Log in
Specifications
Capacity (Metric)
50 L
Material
Cardboard