Learn More
Karl Hecht™ Glass Centrifuge Tubes, Conical
Clear, glass centrifuge tubes provide superior visibility. Karl Hecht™ Glass Centrifuge Tubes, Conical are available graduated/ungraduated, rimmed/rimless, and constructed of AR™ soda glass or Duran™ borosilicate.
Brand:  Karl Hecht™ 40940310
 View more versions of this product
Product Code. 10319161
  • 120.00€ / Pack of 100
Estimated Shipment: 28-10-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Conical bottom.
  • Graduated or ungraduated.
  • Rimmed or rimless.
  • AR™ soda glass, or Duran™ borosilicate.
Specifications
Specifications
10 to 15 mL
Centrifuge
112 mm
17 mm
Rimmed Centrifuge Tube
Borosilicate Glass (3.3)
100/Box
Conical
No