Learn More
BUCHI Glass Drying Flasks
Brand:  BUCHI 002965
Product Code. 10406201
  • 220.80€ / Each
Estimated Shipment: 11-07-2025
Specifications
350 mL
56 mm
Glass
Drying Flask
Product Title
BUCHI Glass Drying Flasks > Capacity: 350mL