Learn More
BUCHI Glass Evaporating Flask
Brand:  BUCHI 047991
Product Code. 12045532
  • 418.62€ / Each
Estimated Shipment: 02-06-2025
Specifications
Glass
Evaporating Flask
1 Pack
4000 mL
Documents
One moment while we fetch your results.
Product Title
BUCHI Glass Evaporating Flask > Capacity: 4000mL