Learn More
Hecht Karl™ Glass Microscope Coverslip
Brand:  Hecht Karl™ 100110
Product Code. 11807093
  • 135.00€ / Pack of 1000
Estimated Shipment: 17-03-2025
Specifications
Coverslip
Circle
10 mm
1000/Pk.
Documents
One moment while we fetch your results.
Product Title
Hecht Karl™ Glass Microscope Coverslip > Thickness: 10mm