Learn More
Karl Hecht™ Glass Mortars
An excellent option for grinding samples and materials, especially for pharmacological applications.
39.00€ - 55.00€
Specifications
Finish | Rough Inner |
---|---|
Product Type | Mortar |
Material | Borosilicate Glass (3.3) |
For Use With (Application) | Grinding materials |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
15641497
|
Karl Hecht™
47028010 |
225 mL |
39.00€
Each |
Estimated Shipment: 23-04-2025
Log in to see stock. |
|||||
15651497
|
Karl Hecht™
47028012 |
400 mL |
55.00€
Each |
Estimated Shipment: 25-02-2025
Log in to see stock. |
|||||
Description
- Rough inside aids with grinding
- With spout for easy pouring
- Manufactured from borosilicate glass 3.3
- Available in multiple capacities
Specifications
Rough Inner | |
Mortar | |
Borosilicate Glass (3.3) | |
Grinding materials |