Learn More
WTW™ Glass Overflow Measuring Flask
Replacement sample bottles and other glassware as well as useful accessories for the expansion of application of measuring systems for different parameters.WTW offers a large assortment of glass ware and other products to replace, supplement and to extend your applications.
Brand:  WTW™ 209030
43.70 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10411602
  • 47.20€ / Each
In Stock
Specifications
22.7 mL
BOD
Glass
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
WTW™ Glass Overflow Measuring Flask > Capacity: 22.7mL