Learn More
BUCHI Glass Receiving Flask
Brand:  BUCHI 047233
Product Code. 12058139
  • 103.39€ / Each
Estimated Shipment: 17-06-2025
Specifications
Glass
Plastic and Glass
125 mL
Receiving Flask
1 Pack
Product Title
BUCHI Glass Receiving Flask > Capacity: 125mL