Learn More
Merck Glass Stirred Cell Cylinder
Brand:  Merck XFUF04711
Product Code. 10776901
  • 334.00€ / Each
Estimated Shipment: 15-04-2025
Specifications
Stirred Cell Cylinder
Glass
75 mL
Documents
One moment while we fetch your results.
Product Title
Merck Glass Stirred Cell Cylinder > Capacity: 75mL