Learn More
Glassy carbon rod, 5mm (0.2in) dia, type 1, Thermo Scientific Chemicals
Widely practiced as an electrode material in electrochemistry, as considerably as for high temperature crucibles and as a part of some prosthetic devices. Glassy carbon electrode (GCE) in aqueous solutions is considered to be an inert electrode for hydronium ion reduction.
254.00€ - 767.00€
Specifications
Chemical Name or Material | Glassy carbon rod |
---|---|
CAS | 7440-44-0 |
Packaging | Tube |
MDL Number | MFCD00133992 |
Solubility Information | Insoluble in almost all the solvents. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11354337
|
Thermo Scientific Alfa Aesar
038011.DJ |
50 mm |
254.00€
50mm |
In Stock
Log in to see stock. |
|||||
11344337
|
Thermo Scientific Alfa Aesar
038011.DM |
100 mm |
454.00€
100mm |
In Stock
Log in to see stock. |
|||||
15441477
|
Thermo Scientific Alfa Aesar
038011.DR |
200 mm |
767.00€
200mm |
Estimated Shipment: 10-10-2024
Log in to see stock. |
|||||
Description
Glassy carbon is widely practiced as an electrode material in electrochemistry, as considerably as for high temperature crucibles and as a part of some prosthetic devices. Glassy carbon electrode (GCE) in aqueous solutions is considered to be an inert electrode for hydronium ion reduction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Glassy carbon rod | |
Tube | |
Insoluble in almost all the solvents. |
7440-44-0 | |
MFCD00133992 |
Safety and Handling
missing translation for 'einecsNumber' : 231-153-3
missing translation for 'rtecsNumber' : FF5250100
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only